Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025354.3 | NP_001020525.2 | 2 - 3 | 259 | 62 | |
GenBank mRNA | BX933464.2 | - | 2 - 3 | 216 | 62 | |
CR352579.1 | - | 2 - 3 | 230 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |