Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145226.2 | NP_001138698.2 | 3 - 4 | 470 | 79 | |
XM_015296218.2 | XP_015151704.1 | 4 - 5 | 543 | 79 | ||
XM_015296220.2 | XP_015151706.1 | 4 - 5 | 1020 | 79 | ||
XM_015296222.2 | XP_015151708.1 | 4 - 5 | 545 | 79 | ||
GenBank mRNA | FM200428.1 | - | 3 - 4 | 470 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |