Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMIM20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145430.1 | NP_001138902.1 | 2 - 3 | 383 | 83 | |
GenBank mRNA | BU307818.1 | - | 2 - 3 | 177 | 83 | |
BX933175.1 | - | 2 - 3 | 449 | 83 | ||
BX935882.1 | - | 2 - 3 | 181 | 83 | ||
CR523170.1 | - | 2 - 3 | 383 | 83 | ||
KR150684.1 | - | 2 - 3 | 192 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |