Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VGLL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145990.1 | NP_001139462.1 | 2 - 3 | 383 | 91 | |
XM_015284465.2 | XP_015139951.1 | 2 - 3 | 477 | 91 | ||
XM_015284466.2 | XP_015139952.1 | 2 - 3 | 477 | 91 | ||
XM_015284467.2 | XP_015139953.1 | 2 - 3 | 477 | 91 | ||
GenBank mRNA | FJ594744.1 | - | 2 - 3 | 383 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |