Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PVALB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159315.1 | NP_001152787.1 | 2 - 3 | 190 | 80 | |
XM_015284568.2 | XP_015140054.1 | 3 - 4 | 392 | 80 | ||
XM_015284574.2 | XP_015140060.1 | 4 - 5 | 593 | 80 | ||
XM_015284580.2 | XP_015140066.1 | 3 - 4 | 261 | 80 | ||
XM_025146984.1 | XP_025002752.1 | 3 - 4 | 370 | 80 | ||
XM_025146985.1 | XP_025002753.1 | 2 - 3 | 200 | 80 | ||
GenBank mRNA | CR389683.1 | - | 3 - 4 | 545 | 80 | |
CR390482.1 | - | 2 - 3 | 293 | 80 | ||
FM994924.1 | - | 2 - 3 | 190 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |