Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHOSPHO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204845.2 | NP_990176.1 | 1 - 2 | 121 | 77 | |
XM_015299434.2 | XP_015154920.1 | 3 - 4 | 365 | 77 | ||
XM_015299435.2 | XP_015154921.1 | 3 - 4 | 330 | 77 | ||
GenBank mRNA | AJ006529.1 | - | 1 - 2 | 125 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |