Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOX11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205187.1 | NP_990518.1 | 1 | 1452 | 94 | |
GenBank mRNA | AB012237.1 | - | 1 | 1299 | 94 | |
U12534.1 | - | 1 | 1452 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon |