Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249972.1 | NP_001236901.1 | 4 - 5 | 484 | 93 | |
GenBank mRNA | AB040543.1 | - | 4 - 5 | 484 | 93 | |
BT090229.1 | - | 5 - 6 | 542 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Glycine max Probe spans exons |