Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001250001.2 | NP_001236930.1 | 5 - 6 | 1365 | 102 | |
GenBank mRNA | AB047475.1 | - | 5 - 6 | 1366 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Glycine max Probe spans exons |