Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001250542.1 | NP_001237471.1 | 1 - 2 | 169 | 110 | |
GenBank mRNA | AF019910.1 | - | 1 - 2 | 169 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Glycine max Probe spans exons |