Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001250643.2 | NP_001237572.1 | 5 - 6 | 952 | 66 | |
GenBank mRNA | AF141602.1 | - | 5 - 6 | 957 | 66 | |
AK244280.1 | - | 5 - 6 | 975 | 66 | ||
AK286711.1 | - | 5 - 6 | 982 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Glycine max Probe spans exons |