Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001250674.1 | NP_001237603.1 | 4 - 5 | 312 | 95 | |
GenBank mRNA | AF180143.1 | - | 4 - 5 | 312 | 95 |
Gene Symbol: LOC100804969 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK244076.1 | - | 4 - 5 | 460 | 95 | |
AK245474.1 | - | 4 - 5 | 459 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Glycine max Probe spans exons |