Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF529300.1 | - | 1 - 2 | 128 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Glycine max Probe spans exons |