Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249183.1 | NP_001236112.1 | 9 - 10 | 1866 | 75 | |
GenBank mRNA | AJ010165.1 | - | 9 - 10 | 1866 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Glycine max Probe spans exons |