Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249662.1 | NP_001236591.1 | 15 - 16 | 2144 | 108 | |
GenBank mRNA | AK245291.1 | - | 15 - 16 | 2206 | 108 | |
AY095999.1 | - | 15 - 16 | 2144 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Glycine max Probe spans exons |