Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001250327.1 | NP_001237256.1 | 3 - 4 | 481 | 80 | |
GenBank mRNA | AY263347.1 | - | 3 - 4 | 481 | 80 | |
FJ014778.1 | - | 2 - 3 | 346 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Protein Kinase Detect Genomic DNA Glycine max Probe spans exons |