Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHYA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001251357.1 | NP_001238286.1 | 3 - 4 | 3197 | 89 | |
GenBank mRNA | EU428747.2 | - | 3 - 4 | 3194 | 89 | |
L34844.1 | - | 3 - 4 | 3197 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Protein Kinase 3' Most Detect Genomic DNA Recommended Assay Glycine max Probe spans exons |