Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AM158274.1 | - | 13 - 14 | 1668 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Glycine max Probe spans exons |