Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249446.1 | NP_001236375.1 | 3 - 4 | 985 | 70 | |
GenBank mRNA | AJ875407.1 | - | 3 - 4 | 985 | 70 | |
AK245571.1 | - | 3 - 4 | 1002 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Glycine max Probe spans exons |