Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AM085508.1 | - | 7 - 8 | 1402 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Glycine max Probe spans exons |