Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001251288.1 | NP_001238217.1 | 19 - 20 | 2277 | 65 | |
GenBank mRNA | AY856369.1 | - | 19 - 20 | 2277 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Glycine max Probe spans exons |