Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001248095.1 | NP_001235024.1 | 3 - 4 | 405 | 102 | |
GenBank mRNA | EF521873.1 | - | 3 - 4 | 405 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Glycine max Probe spans exons |