Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK245933.1 | - | 3 - 4 | 497 | 82 | |
EU019565.1 | - | 3 - 4 | 474 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Glycine max Probe spans exons |