Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001250709.1 | NP_001237638.1 | 1 - 2 | 520 | 101 | |
GenBank mRNA | EU375354.1 | - | 1 - 2 | 520 | 101 |
Gene Symbol: LOC100778162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK244967.1 | - | 1 - 2 | 425 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Glycine max Probe spans exons |