Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249042.1 | NP_001235971.1 | 8 - 9 | 2002 | 79 | |
GenBank mRNA | FJ795039.1 | - | 8 - 9 | 2002 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Protein Kinase 3' Most Detect Genomic DNA Glycine max Probe spans exons |