Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHI1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249826.2 | NP_001236755.1 | 4 | 750 | 100 | |
GenBank mRNA | AY595414.1 | - | 4 | 753 | 100 | |
BT095642.1 | - | 4 | 752 | 100 | ||
DQ191404.1 | - | 4 | 728 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Glycine max Both primers and probe map within a single exon |