Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHI1B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249168.1 | NP_001236097.1 | 4 | 653 | 98 | |
GenBank mRNA | AY595419.1 | - | 4 | 653 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Glycine max Both primers and probe map within a single exon |