Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BZIP49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | DQ787017.1 | - | 1 | 145 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Glycine max Both primers and probe map within a single exon |