Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001250848.1 | NP_001237777.1 | 3 | 943 | 106 | |
GenBank mRNA | EU019572.1 | - | 3 | 943 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Glycine max Both primers and probe map within a single exon |