Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006830.hg.1 |
ENST00000239882.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006830.hg.1 |
ENST00000442101.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006830.hg.1 |
ENST00000498824.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006830.hg.1 |
NM_001145353.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006830.hg.1 |
NM_172373.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006830.hg.1 |
TC13000597.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000239882.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000239882.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000239882.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000239882.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000239882.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000239882.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000239882.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000239882.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000239882.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000239882.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000239882.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000239882.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000239882.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000239882.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000405737.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000405737.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000405737.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000405737.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000405737.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000442101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000442101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000442101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000442101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000442101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000442101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000442101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000442101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000442101.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000498824.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000498824.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000498824.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000498824.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000498824.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000498824.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000498824.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000498824.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000498824.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000498824.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000498824.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000498824.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000498824.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000625359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000625359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000625359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000625359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000625359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000625359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000625359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000625359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000625359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
ENST00000625359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
NM_001145353.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
NM_001145353.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
NM_001145353.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
NM_001145353.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
NM_001145353.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
NM_001145353.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
NM_001145353.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
NM_001145353.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
NM_001145353.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
NM_001145353.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
NM_001145353.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
NM_001145353.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
NM_001145353.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
NM_172373.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
NM_172373.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
NM_172373.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
NM_172373.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
NM_172373.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
NM_172373.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
NM_172373.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
NM_172373.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
NM_172373.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
NM_172373.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
NM_172373.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
NM_172373.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
NM_172373.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
NM_172373.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
NM_172373.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008702.hg.1 |
NM_172373.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158782.hg.1 |
ENST00000239882.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158782.hg.1 |
ENST00000405737.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158782.hg.1 |
ENST00000498824.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158782.hg.1 |
ENST00000625359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158782.hg.1 |
NM_001145353.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158782.hg.1 |
NM_172373.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158782.hg.1 |
ENST00000239882.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158782.hg.1 |
ENST00000405737.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158782.hg.1 |
ENST00000498824.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158782.hg.1 |
ENST00000625359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158782.hg.1 |
NM_001145353.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158782.hg.1 |
NM_172373.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158782.hg.1 |
TC1300008702.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300158781.hg.1 |
ENST00000239882.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158781.hg.1 |
ENST00000498824.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158781.hg.1 |
ENST00000625359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158781.hg.1 |
NM_001145353.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158781.hg.1 |
NM_172373.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158781.hg.1 |
ENST00000239882.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158781.hg.1 |
ENST00000498824.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158781.hg.1 |
ENST00000625359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158781.hg.1 |
NM_001145353.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158781.hg.1 |
NM_172373.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158781.hg.1 |
TC1300008702.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000239882.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000239882.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000239882.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000239882.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000239882.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000239882.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000239882.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000239882.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000239882.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000239882.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000239882.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000239882.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000239882.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000239882.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000405737.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000405737.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000405737.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000405737.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000405737.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000442101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000442101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000442101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000442101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000442101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000442101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000442101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000442101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000442101.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000498824.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000498824.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000498824.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000498824.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000498824.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000498824.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000498824.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000498824.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000498824.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000498824.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000498824.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000498824.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000498824.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000625359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000625359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000625359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000625359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000625359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000625359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000625359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000625359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000625359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
ENST00000625359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
NM_001145353.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
NM_001145353.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
NM_001145353.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
NM_001145353.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
NM_001145353.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
NM_001145353.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
NM_001145353.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
NM_001145353.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
NM_001145353.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
NM_001145353.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
NM_001145353.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
NM_001145353.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
NM_001145353.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
NM_172373.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
NM_172373.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
NM_172373.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
NM_172373.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
NM_172373.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
NM_172373.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
NM_172373.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
NM_172373.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
NM_172373.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
NM_172373.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
NM_172373.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
NM_172373.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
NM_172373.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
NM_172373.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
NM_172373.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008702.hg.1 |
NM_172373.3 |
Refseq |