Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318789.1 | NP_001305718.1 | 2 - 3 | 256 | 80 | |
NM_001318790.1 | NP_001305719.1 | 2 - 3 | 233 | 80 | ||
NM_001318791.1 | NP_001305720.1 | 2 - 3 | 222 | 80 | ||
NM_001318793.1 | NP_001305722.1 | 2 - 3 | 218 | 80 | ||
NM_003264.4 | NP_003255.2 | 2 - 3 | 241 | 80 | ||
GenBank mRNA | AF051152.1 | - | 1 - 2 | 147 | 80 | |
BI769109.1 | - | 2 - 3 | 204 | 80 | ||
DA460298.1 | - | 3 - 4 | 207 | 80 | ||
U88878.1 | - | 1 - 2 | 114 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Human NFkB Pathway Inventoried Human_Nfkb Probe spans exons |