Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010910.hg.1 |
ENST00000398189.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010910.hg.1 |
ENST00000398189.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010910.hg.1 |
ENST00000398189.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010910.hg.1 |
NM_001638.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010910.hg.1 |
NM_001638.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010910.hg.1 |
NM_001638.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12021416.hg.1 |
ENST00000398189.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12021416.hg.1 |
ENST00000541105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12021416.hg.1 |
NM_001638.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12021416.hg.1 |
TC12001604.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12021417.hg.1 |
ENST00000398189.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12021417.hg.1 |
ENST00000541105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12021417.hg.1 |
NM_001638.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12021417.hg.1 |
TC12001604.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1200010910.hg.1 |
ENST00000398189.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010910.hg.1 |
ENST00000398189.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010910.hg.1 |
ENST00000398189.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010910.hg.1 |
NM_001638.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010910.hg.1 |
NM_001638.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010910.hg.1 |
NM_001638.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200184298.hg.1 |
ENST00000398189.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200184298.hg.1 |
NM_001638.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200184298.hg.1 |
ENST00000398189.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200184298.hg.1 |
NM_001638.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200184298.hg.1 |
TC1200010910.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200184296.hg.1 |
ENST00000398189.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200184296.hg.1 |
NM_001638.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200184296.hg.1 |
ENST00000398189.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200184296.hg.1 |
NM_001638.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200184296.hg.1 |
TC1200010910.hg.1 |
Clariom D TC |