Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025109.1 | NP_001020280.1 | 4 - 5 | 854 | 63 | |
NM_001773.2 | NP_001764.1 | 4 - 5 | 854 | 63 | ||
GenBank mRNA | AF202879.1 | - | 2 - 3 | 99 | 63 | |
AF523361.1 | - | 4 - 5 | 667 | 63 | ||
AF538601.1 | - | 3 - 4 | 505 | 63 | ||
AK291529.1 | - | 4 - 5 | 607 | 63 | ||
AK293030.1 | - | 4 - 5 | 631 | 63 | ||
AK294381.1 | - | 3 - 4 | 378 | 63 | ||
AK300835.1 | - | 5 - 6 | 671 | 63 | ||
BC039146.1 | - | 4 - 5 | 604 | 4746591 | 63 | |
M81104.1 | - | 4 - 5 | 853 | 63 | ||
S53910.1 | - | 4 - 5 | 686 | 63 | ||
S53911.1 | - | 4 - 5 | 686 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Immune Tolerance Network Mammalian Gene Collection Homo sapiens Human Stem Cell Pluripotency Human Immune Response Inventoried Human_Immune Human_Stem_Cell Probe spans exons |