Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011564.hg.1 |
ENST00000266003.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011564.hg.1 |
ENST00000266003.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011564.hg.1 |
ENST00000266003.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011564.hg.1 |
ENST00000266003.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011564.hg.1 |
ENST00000266003.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011564.hg.1 |
ENST00000266003.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011564.hg.1 |
ENST00000430124.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011564.hg.1 |
ENST00000430124.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011564.hg.1 |
ENST00000430124.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011564.hg.1 |
ENST00000430124.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011564.hg.1 |
ENST00000430124.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011564.hg.1 |
ENST00000430124.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011564.hg.1 |
ENST00000507738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011564.hg.1 |
ENST00000507738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011564.hg.1 |
ENST00000507738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011564.hg.1 |
ENST00000507738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011564.hg.1 |
ENST00000507738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011564.hg.1 |
NM_001040109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011564.hg.1 |
NM_001040109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011564.hg.1 |
NM_001040109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011564.hg.1 |
NM_001040109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011564.hg.1 |
NM_001040109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011564.hg.1 |
NM_001040109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011564.hg.1 |
NM_001184698.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011564.hg.1 |
NM_001184698.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011564.hg.1 |
NM_001184698.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011564.hg.1 |
NM_001184698.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011564.hg.1 |
NM_001184698.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011564.hg.1 |
NM_001184698.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011564.hg.1 |
NM_002418.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011564.hg.1 |
NM_002418.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011564.hg.1 |
NM_002418.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011564.hg.1 |
NM_002418.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011564.hg.1 |
NM_002418.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011564.hg.1 |
NM_002418.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06019424.hg.1 |
TC06001594.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06019424.hg.1 |
ENST00000266003.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06019424.hg.1 |
ENST00000430124.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06019424.hg.1 |
ENST00000507738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06019424.hg.1 |
NM_001040109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06019424.hg.1 |
NM_001184698.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06019424.hg.1 |
NM_002418.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06019425.hg.1 |
TC06001594.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06019425.hg.1 |
ENST00000266003.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06019425.hg.1 |
ENST00000430124.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06019425.hg.1 |
ENST00000507738.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06019425.hg.1 |
NM_001040109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06019425.hg.1 |
NM_001184698.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06019425.hg.1 |
NM_002418.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011564.hg.1 |
ENST00000266003.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011564.hg.1 |
ENST00000266003.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011564.hg.1 |
ENST00000266003.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011564.hg.1 |
ENST00000266003.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011564.hg.1 |
ENST00000266003.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011564.hg.1 |
ENST00000266003.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011564.hg.1 |
ENST00000430124.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011564.hg.1 |
ENST00000430124.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011564.hg.1 |
ENST00000430124.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011564.hg.1 |
ENST00000430124.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011564.hg.1 |
ENST00000430124.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011564.hg.1 |
ENST00000430124.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011564.hg.1 |
ENST00000507738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011564.hg.1 |
ENST00000507738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011564.hg.1 |
ENST00000507738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011564.hg.1 |
ENST00000507738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011564.hg.1 |
ENST00000507738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011564.hg.1 |
NM_001040109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011564.hg.1 |
NM_001040109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011564.hg.1 |
NM_001040109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011564.hg.1 |
NM_001040109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011564.hg.1 |
NM_001040109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011564.hg.1 |
NM_001040109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011564.hg.1 |
NM_001184698.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011564.hg.1 |
NM_001184698.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011564.hg.1 |
NM_001184698.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011564.hg.1 |
NM_001184698.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011564.hg.1 |
NM_001184698.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011564.hg.1 |
NM_001184698.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011564.hg.1 |
NM_002418.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011564.hg.1 |
NM_002418.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011564.hg.1 |
NM_002418.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011564.hg.1 |
NM_002418.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011564.hg.1 |
NM_002418.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011564.hg.1 |
NM_002418.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600179296.hg.1 |
ENST00000266003.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600179296.hg.1 |
ENST00000430124.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600179296.hg.1 |
ENST00000507738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600179296.hg.1 |
NM_001040109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600179296.hg.1 |
NM_001184698.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600179296.hg.1 |
NM_002418.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600179296.hg.1 |
ENST00000266003.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600179296.hg.1 |
ENST00000430124.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600179296.hg.1 |
ENST00000507738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600179296.hg.1 |
NM_001040109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600179296.hg.1 |
NM_001184698.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600179296.hg.1 |
NM_002418.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600179296.hg.1 |
TC0600011564.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600179294.hg.1 |
ENST00000266003.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600179294.hg.1 |
ENST00000430124.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600179294.hg.1 |
ENST00000507738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600179294.hg.1 |
NM_001040109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600179294.hg.1 |
NM_001184698.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600179294.hg.1 |
NM_002418.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600179294.hg.1 |
ENST00000266003.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600179294.hg.1 |
ENST00000430124.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600179294.hg.1 |
ENST00000507738.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600179294.hg.1 |
NM_001040109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600179294.hg.1 |
NM_001184698.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600179294.hg.1 |
NM_002418.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600179294.hg.1 |
TC0600011564.hg.1 |
Clariom D TC |