Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000164.hg.1 |
ENST00000234347.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000164.hg.1 |
ENST00000544537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000164.hg.1 |
NM_002777.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000164.hg.1 |
TC19000018.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006492.hg.1 |
ENST00000234347.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006492.hg.1 |
ENST00000234347.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006492.hg.1 |
ENST00000234347.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006492.hg.1 |
ENST00000234347.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006492.hg.1 |
ENST00000234347.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006492.hg.1 |
ENST00000234347.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006492.hg.1 |
ENST00000234347.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006492.hg.1 |
ENST00000234347.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006492.hg.1 |
ENST00000544537.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006492.hg.1 |
ENST00000544537.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006492.hg.1 |
ENST00000544537.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006492.hg.1 |
ENST00000544537.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006492.hg.1 |
ENST00000544537.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006492.hg.1 |
ENST00000544537.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006492.hg.1 |
ENST00000544537.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006492.hg.1 |
ENST00000544537.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006492.hg.1 |
NM_002777.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006492.hg.1 |
NM_002777.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006492.hg.1 |
NM_002777.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006492.hg.1 |
NM_002777.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006492.hg.1 |
NM_002777.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006492.hg.1 |
NM_002777.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006492.hg.1 |
NM_002777.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006492.hg.1 |
NM_002777.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000162.hg.1 |
ENST00000234347.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000162.hg.1 |
NM_002777.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000162.hg.1 |
TC19000018.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900144540.hg.1 |
ENST00000234347.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144540.hg.1 |
ENST00000544537.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144540.hg.1 |
NM_002777.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900144540.hg.1 |
TC1900006492.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900144540.hg.1 |
ENST00000234347.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144540.hg.1 |
ENST00000544537.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144540.hg.1 |
NM_002777.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006492.hg.1 |
ENST00000234347.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006492.hg.1 |
ENST00000234347.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006492.hg.1 |
ENST00000234347.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006492.hg.1 |
ENST00000234347.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006492.hg.1 |
ENST00000234347.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006492.hg.1 |
ENST00000234347.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006492.hg.1 |
ENST00000234347.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006492.hg.1 |
ENST00000234347.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006492.hg.1 |
ENST00000544537.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006492.hg.1 |
ENST00000544537.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006492.hg.1 |
ENST00000544537.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006492.hg.1 |
ENST00000544537.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006492.hg.1 |
ENST00000544537.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006492.hg.1 |
ENST00000544537.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006492.hg.1 |
ENST00000544537.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006492.hg.1 |
ENST00000544537.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006492.hg.1 |
NM_002777.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006492.hg.1 |
NM_002777.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006492.hg.1 |
NM_002777.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006492.hg.1 |
NM_002777.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006492.hg.1 |
NM_002777.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006492.hg.1 |
NM_002777.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006492.hg.1 |
NM_002777.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006492.hg.1 |
NM_002777.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900144537.hg.1 |
ENST00000234347.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144537.hg.1 |
NM_002777.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900144537.hg.1 |
TC1900006492.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900144537.hg.1 |
ENST00000234347.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144537.hg.1 |
NM_002777.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900144541.hg.1 |
ENST00000234347.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144541.hg.1 |
ENST00000544537.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144541.hg.1 |
NM_002777.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900144541.hg.1 |
TC1900006492.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900144541.hg.1 |
ENST00000234347.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144541.hg.1 |
ENST00000544537.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144541.hg.1 |
NM_002777.3 |
Refseq |