Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004558.hg.1 |
ENST00000266088.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004558.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004558.hg.1 |
TC22000237.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004559.hg.1 |
ENST00000266088.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004559.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004559.hg.1 |
TC22000237.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000477969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000477969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000477969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000477969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000477969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000477969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000486394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000486394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000486394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000486394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000543737.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000543737.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000543737.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000543737.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000543737.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000543737.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000543737.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000543737.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000543737.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000543737.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000543737.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000543737.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000543737.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000543737.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000543737.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
ENST00000543737.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
NM_001256314.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
NM_001256314.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
NM_001256314.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
NM_001256314.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
NM_001256314.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
NM_001256314.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
NM_001256314.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
NM_001256314.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
NM_001256314.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
NM_001256314.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
NM_001256314.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
NM_001256314.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
NM_001256314.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
NM_001256314.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
NM_001256314.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007120.hg.1 |
NM_001256314.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004556.hg.1 |
ENST00000266088.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004556.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004556.hg.1 |
TC22000237.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004562.hg.1 |
ENST00000266088.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004562.hg.1 |
ENST00000543737.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004562.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004562.hg.1 |
NM_001256314.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004562.hg.1 |
TC22000237.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2200150165.hg.1 |
TC2200007120.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200150165.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150165.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200150165.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150165.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200150160.hg.1 |
TC2200007120.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200150160.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150160.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200150160.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150160.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200150168.hg.1 |
TC2200007120.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200150168.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150168.hg.1 |
ENST00000543737.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150168.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200150168.hg.1 |
NM_001256314.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200150168.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150168.hg.1 |
ENST00000543737.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150168.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200150168.hg.1 |
NM_001256314.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000477969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000477969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000477969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000477969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000477969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000477969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000486394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000486394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000486394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000486394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000543737.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000543737.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000543737.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000543737.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000543737.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000543737.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000543737.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000543737.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000543737.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000543737.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000543737.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000543737.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000543737.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000543737.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000543737.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
ENST00000543737.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
NM_001256314.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
NM_001256314.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
NM_001256314.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
NM_001256314.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
NM_001256314.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
NM_001256314.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
NM_001256314.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
NM_001256314.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
NM_001256314.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
NM_001256314.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
NM_001256314.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
NM_001256314.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
NM_001256314.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
NM_001256314.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
NM_001256314.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007120.hg.1 |
NM_001256314.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200150163.hg.1 |
TC2200007120.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200150163.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150163.hg.1 |
NM_000343.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200150163.hg.1 |
ENST00000266088.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150163.hg.1 |
NM_000343.3 |
Refseq |