Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007309.hg.1 |
TC07000519.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007309.hg.1 |
ENST00000361669.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007309.hg.1 |
ENST00000394720.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007309.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007309.hg.1 |
ENST00000536043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007309.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007310.hg.1 |
TC07000519.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007310.hg.1 |
ENST00000361669.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007310.hg.1 |
ENST00000536043.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007310.hg.1 |
ENST00000546348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007310.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000421579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000421579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000421579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000421579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000441140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000441140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000441140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000454217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000454217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000454217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
ENST00000454217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000421579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000421579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000421579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000421579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000441140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000441140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000441140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000454217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000454217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000454217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
ENST00000454217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008240.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157164.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157164.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157164.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157164.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157164.hg.1 |
ENST00000439827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157164.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157164.hg.1 |
TC0700008240.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700157165.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157165.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157165.hg.1 |
ENST00000361669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700157165.hg.1 |
NM_000840.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700157165.hg.1 |
TC0700008240.hg.1 |
Clariom D TC |