Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
ENST00000215909.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
ENST00000215909.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
ENST00000215909.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
ENST00000215909.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
ENST00000215909.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
ENST00000215909.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
ENST00000215909.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
ENST00000425542.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
ENST00000425542.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
ENST00000425542.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
ENST00000425542.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
ENST00000425542.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
ENST00000425542.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
ENST00000425542.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
ENST00000454173.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
ENST00000454173.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
ENST00000454173.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
ENST00000464120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
ENST00000464120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
ENST00000464120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
ENST00000472321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
ENST00000472321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
ENST00000472321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
ENST00000472321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
ENST00000472321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
ENST00000472321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
ENST00000480207.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
ENST00000480207.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
ENST00000489315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
ENST00000489315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
ENST00000489315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
ENST00000489315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
ENST00000489315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
ENST00000489315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
NM_002305.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
NM_002305.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
NM_002305.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
NM_002305.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
NM_002305.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
NM_002305.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007312.hg.1 |
NM_002305.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005276.hg.1 |
ENST00000215909.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005276.hg.1 |
NM_002305.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005276.hg.1 |
TC22000285.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005277.hg.1 |
ENST00000215909.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005277.hg.1 |
NM_002305.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005277.hg.1 |
TC22000285.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005279.hg.1 |
ENST00000215909.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005279.hg.1 |
NM_002305.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005279.hg.1 |
TC22000285.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005284.hg.1 |
ENST00000215909.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005284.hg.1 |
ENST00000489315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005284.hg.1 |
NM_002305.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005284.hg.1 |
TC22000285.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005278.hg.1 |
ENST00000215909.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005278.hg.1 |
NM_002305.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005278.hg.1 |
TC22000285.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005280.hg.1 |
ENST00000215909.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005280.hg.1 |
NM_002305.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005280.hg.1 |
TC22000285.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2200151310.hg.1 |
TC2200007312.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200151310.hg.1 |
ENST00000215909.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151310.hg.1 |
NM_002305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200151310.hg.1 |
ENST00000215909.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151310.hg.1 |
NM_002305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
ENST00000215909.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
ENST00000215909.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
ENST00000215909.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
ENST00000215909.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
ENST00000215909.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
ENST00000215909.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
ENST00000215909.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
ENST00000425542.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
ENST00000425542.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
ENST00000425542.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
ENST00000425542.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
ENST00000425542.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
ENST00000425542.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
ENST00000425542.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
ENST00000454173.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
ENST00000454173.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
ENST00000454173.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
ENST00000464120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
ENST00000464120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
ENST00000464120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
ENST00000472321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
ENST00000472321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
ENST00000472321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
ENST00000472321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
ENST00000472321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
ENST00000472321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
ENST00000480207.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
ENST00000480207.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
ENST00000489315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
ENST00000489315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
ENST00000489315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
ENST00000489315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
ENST00000489315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
ENST00000489315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
NM_002305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
NM_002305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
NM_002305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
NM_002305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
NM_002305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
NM_002305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007312.hg.1 |
NM_002305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200151315.hg.1 |
TC2200007312.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200151315.hg.1 |
ENST00000215909.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151315.hg.1 |
ENST00000425542.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151315.hg.1 |
ENST00000464120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151315.hg.1 |
NM_002305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200151315.hg.1 |
ENST00000215909.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151315.hg.1 |
ENST00000425542.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151315.hg.1 |
ENST00000464120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151315.hg.1 |
NM_002305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200151323.hg.1 |
TC2200007312.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200151323.hg.1 |
ENST00000215909.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151323.hg.1 |
ENST00000425542.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151323.hg.1 |
ENST00000454173.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151323.hg.1 |
ENST00000464120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151323.hg.1 |
ENST00000472321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151323.hg.1 |
ENST00000480207.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151323.hg.1 |
ENST00000489315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151323.hg.1 |
NM_002305.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200151323.hg.1 |
ENST00000215909.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151323.hg.1 |
ENST00000425542.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151323.hg.1 |
ENST00000454173.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151323.hg.1 |
ENST00000464120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151323.hg.1 |
ENST00000472321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151323.hg.1 |
ENST00000480207.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151323.hg.1 |
ENST00000489315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151323.hg.1 |
NM_002305.3 |
Refseq |