Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018898.3 | NP_061721.2 | 1 - 2 | 2613 | 114 | |
GenBank mRNA | AF152303.1 | - | 1 - 2 | 2437 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Inventoried Probe spans exons |