Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000294728.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000294728.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000294728.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000294728.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000294728.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000294728.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000294728.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000294728.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000294728.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000294728.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000294728.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000294728.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000294728.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000347652.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000347652.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000347652.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000347652.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000347652.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000347652.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000347652.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000347652.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000347652.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000347652.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000347652.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000347652.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000370115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000370115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000370115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000370115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000370115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000370115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000370115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000370115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000370115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000370115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000370115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000370119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000370119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000370119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000370119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000370119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000370119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000370119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000370119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000370119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000370119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000370119.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000603679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000603679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000603679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
ENST00000603679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_001078.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_001078.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_001078.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_001078.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_001078.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_001078.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_001078.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_001078.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_001078.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_001078.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_001078.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_001078.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_001078.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_001078.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_001199834.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_001199834.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_001199834.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_001199834.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_001199834.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_001199834.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_001199834.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_001199834.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_001199834.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_001199834.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_001199834.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_001199834.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_001199834.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_080682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_080682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_080682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_080682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_080682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_080682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_080682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_080682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_080682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_080682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_080682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_080682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009221.hg.1 |
NM_080682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014255.hg.1 |
TC01000904.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014255.hg.1 |
ENST00000294728.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014255.hg.1 |
ENST00000347652.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014255.hg.1 |
ENST00000370119.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014255.hg.1 |
NM_001078.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014255.hg.1 |
NM_001199834.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014255.hg.1 |
NM_080682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000294728.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000294728.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000294728.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000294728.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000294728.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000294728.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000294728.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000294728.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000294728.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000294728.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000294728.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000294728.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000294728.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000347652.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000347652.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000347652.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000347652.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000347652.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000347652.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000347652.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000347652.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000347652.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000347652.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000347652.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000347652.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000370115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000370115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000370115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000370115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000370115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000370115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000370115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000370115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000370115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000370115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000370115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000370119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000370119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000370119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000370119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000370119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000370119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000370119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000370119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000370119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000370119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000370119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000603679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000603679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000603679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
ENST00000603679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_001078.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_001078.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_001078.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_001078.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_001078.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_001078.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_001078.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_001078.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_001078.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_001078.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_001078.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_001078.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_001078.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_001078.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_001199834.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_001199834.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_001199834.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_001199834.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_001199834.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_001199834.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_001199834.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_001199834.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_001199834.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_001199834.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_001199834.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_001199834.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_001199834.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_080682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_080682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_080682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_080682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_080682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_080682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_080682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_080682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_080682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_080682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_080682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_080682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009221.hg.1 |
NM_080682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165690.hg.1 |
ENST00000294728.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165690.hg.1 |
ENST00000347652.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165690.hg.1 |
ENST00000370119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165690.hg.1 |
NM_001078.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165690.hg.1 |
NM_001199834.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165690.hg.1 |
NM_080682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165690.hg.1 |
ENST00000294728.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165690.hg.1 |
ENST00000347652.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165690.hg.1 |
ENST00000370119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165690.hg.1 |
NM_001078.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165690.hg.1 |
NM_001199834.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165690.hg.1 |
NM_080682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165690.hg.1 |
TC0100009221.hg.1 |
Clariom D TC |