Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000756.3 | NP_000747.1 | 1 - 2 | 355 | 131 | |
XM_017013090.1 | XP_016868579.1 | 5 - 6 | 693 | 131 | ||
XM_017013091.1 | XP_016868580.1 | 5 - 6 | 756 | 131 | ||
XM_017013092.1 | XP_016868581.1 | 5 - 6 | 694 | 131 | ||
XM_017013093.1 | XP_016868582.1 | 5 - 6 | 833 | 131 | ||
GenBank mRNA | AK075431.1 | - | 1 - 2 | 301 | 131 | |
BC002599.2 | - | 1 - 2 | 166 | 3160933 | 131 | |
CB992265.1 | - | 1 - 2 | 382 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried MicroArray Quality Control (MAQC) Probe spans exons |