Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000543.hg.1 |
ENST00000245983.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000543.hg.1 |
ENST00000359100.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000543.hg.1 |
ENST00000359987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000543.hg.1 |
ENST00000380346.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000543.hg.1 |
ENST00000380347.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000543.hg.1 |
NM_001501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000543.hg.1 |
NM_178331.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000543.hg.1 |
NM_178332.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000543.hg.1 |
TC20000031.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000539.hg.1 |
ENST00000245983.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000539.hg.1 |
ENST00000359100.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000539.hg.1 |
ENST00000359987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000539.hg.1 |
NM_001501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000539.hg.1 |
NM_178331.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000539.hg.1 |
NM_178332.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000539.hg.1 |
TC20000031.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
ENST00000245983.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
ENST00000245983.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
ENST00000245983.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
ENST00000245983.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
ENST00000245983.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
ENST00000359100.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
ENST00000359100.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
ENST00000359100.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
ENST00000359100.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
ENST00000359100.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
ENST00000380346.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
ENST00000380346.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
ENST00000380346.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
ENST00000380346.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
ENST00000380347.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
ENST00000380347.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
ENST00000380347.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
ENST00000380347.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
ENST00000380347.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
NM_001310220.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
NM_001310220.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
NM_001310220.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
NM_001310220.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
NM_001310220.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
NM_001501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
NM_001501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
NM_001501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
NM_001501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
NM_001501.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
NM_178331.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
NM_178331.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
NM_178331.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
NM_178331.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
NM_178331.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
NM_178332.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
NM_178332.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
NM_178332.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
NM_178332.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006533.hg.1 |
NM_178332.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000144894.hg.1 |
ENST00000245983.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000144894.hg.1 |
ENST00000359100.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000144894.hg.1 |
NM_001310220.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000144894.hg.1 |
NM_001501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000144894.hg.1 |
NM_178331.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000144894.hg.1 |
NM_178332.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000144894.hg.1 |
TC2000006533.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000144894.hg.1 |
ENST00000245983.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000144894.hg.1 |
ENST00000359100.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000144894.hg.1 |
NM_001310220.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000144894.hg.1 |
NM_001501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000144894.hg.1 |
NM_178331.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000144894.hg.1 |
NM_178332.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000144899.hg.1 |
ENST00000245983.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000144899.hg.1 |
ENST00000359100.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000144899.hg.1 |
ENST00000380346.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000144899.hg.1 |
ENST00000380347.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000144899.hg.1 |
NM_001310220.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000144899.hg.1 |
NM_001501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000144899.hg.1 |
NM_178331.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000144899.hg.1 |
NM_178332.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000144899.hg.1 |
TC2000006533.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000144899.hg.1 |
ENST00000245983.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000144899.hg.1 |
ENST00000359100.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000144899.hg.1 |
ENST00000380346.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000144899.hg.1 |
ENST00000380347.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000144899.hg.1 |
NM_001310220.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000144899.hg.1 |
NM_001501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000144899.hg.1 |
NM_178331.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000144899.hg.1 |
NM_178332.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
ENST00000245983.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
ENST00000245983.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
ENST00000245983.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
ENST00000245983.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
ENST00000245983.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
ENST00000359100.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
ENST00000359100.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
ENST00000359100.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
ENST00000359100.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
ENST00000359100.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
ENST00000380346.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
ENST00000380346.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
ENST00000380346.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
ENST00000380346.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
ENST00000380347.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
ENST00000380347.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
ENST00000380347.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
ENST00000380347.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
ENST00000380347.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
NM_001310220.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
NM_001310220.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
NM_001310220.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
NM_001310220.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
NM_001310220.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
NM_001501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
NM_001501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
NM_001501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
NM_001501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
NM_001501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
NM_178331.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
NM_178331.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
NM_178331.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
NM_178331.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
NM_178331.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
NM_178332.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
NM_178332.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
NM_178332.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
NM_178332.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006533.hg.1 |
NM_178332.1 |
Refseq |