Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05028492.hg.1 |
TC05002075.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05028492.hg.1 |
ENST00000265087.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05028492.hg.1 |
NM_003714.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05028489.hg.1 |
TC05002075.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05028489.hg.1 |
ENST00000265087.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05028489.hg.1 |
ENST00000520593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05028489.hg.1 |
NM_003714.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05028496.hg.1 |
TC05002075.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05028496.hg.1 |
ENST00000265087.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05028496.hg.1 |
NM_003714.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012870.hg.1 |
ENST00000265087.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012870.hg.1 |
ENST00000265087.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012870.hg.1 |
ENST00000265087.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012870.hg.1 |
ENST00000265087.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012870.hg.1 |
ENST00000265087.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012870.hg.1 |
ENST00000265087.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012870.hg.1 |
ENST00000265087.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012870.hg.1 |
ENST00000265087.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012870.hg.1 |
ENST00000265087.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012870.hg.1 |
ENST00000265087.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012870.hg.1 |
ENST00000265087.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012870.hg.1 |
ENST00000518455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012870.hg.1 |
ENST00000518455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012870.hg.1 |
ENST00000518455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012870.hg.1 |
ENST00000519511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012870.hg.1 |
ENST00000519511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012870.hg.1 |
ENST00000519511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012870.hg.1 |
ENST00000519511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012870.hg.1 |
ENST00000519511.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012870.hg.1 |
ENST00000520593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012870.hg.1 |
ENST00000520593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012870.hg.1 |
ENST00000520593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012870.hg.1 |
ENST00000520593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012870.hg.1 |
ENST00000520648.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012870.hg.1 |
ENST00000520648.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012870.hg.1 |
ENST00000520648.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012870.hg.1 |
ENST00000520648.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012870.hg.1 |
NM_003714.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012870.hg.1 |
NM_003714.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012870.hg.1 |
NM_003714.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012870.hg.1 |
NM_003714.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012870.hg.1 |
NM_003714.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012870.hg.1 |
NM_003714.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012870.hg.1 |
NM_003714.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012870.hg.1 |
NM_003714.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012870.hg.1 |
NM_003714.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012870.hg.1 |
NM_003714.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012870.hg.1 |
NM_003714.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500189429.hg.1 |
ENST00000265087.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189429.hg.1 |
ENST00000518455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189429.hg.1 |
ENST00000519511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189429.hg.1 |
ENST00000520648.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189429.hg.1 |
NM_003714.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500189429.hg.1 |
ENST00000265087.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189429.hg.1 |
ENST00000518455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189429.hg.1 |
ENST00000519511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189429.hg.1 |
ENST00000520648.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189429.hg.1 |
NM_003714.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500189429.hg.1 |
TC0500012870.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500189433.hg.1 |
ENST00000265087.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189433.hg.1 |
ENST00000519511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189433.hg.1 |
NM_003714.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500189433.hg.1 |
ENST00000265087.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189433.hg.1 |
ENST00000519511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189433.hg.1 |
NM_003714.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500189433.hg.1 |
TC0500012870.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500189426.hg.1 |
ENST00000265087.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189426.hg.1 |
ENST00000518455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189426.hg.1 |
ENST00000520593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189426.hg.1 |
ENST00000520648.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189426.hg.1 |
NM_003714.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500189426.hg.1 |
ENST00000265087.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189426.hg.1 |
ENST00000518455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189426.hg.1 |
ENST00000520593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189426.hg.1 |
ENST00000520648.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189426.hg.1 |
NM_003714.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500189426.hg.1 |
TC0500012870.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500189432.hg.1 |
ENST00000265087.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189432.hg.1 |
ENST00000519511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189432.hg.1 |
NM_003714.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500189432.hg.1 |
ENST00000265087.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189432.hg.1 |
ENST00000519511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500189432.hg.1 |
NM_003714.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500189432.hg.1 |
TC0500012870.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500012870.hg.1 |
ENST00000265087.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012870.hg.1 |
ENST00000265087.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012870.hg.1 |
ENST00000265087.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012870.hg.1 |
ENST00000265087.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012870.hg.1 |
ENST00000265087.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012870.hg.1 |
ENST00000265087.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012870.hg.1 |
ENST00000265087.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012870.hg.1 |
ENST00000265087.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012870.hg.1 |
ENST00000265087.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012870.hg.1 |
ENST00000265087.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012870.hg.1 |
ENST00000265087.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012870.hg.1 |
ENST00000518455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012870.hg.1 |
ENST00000518455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012870.hg.1 |
ENST00000518455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012870.hg.1 |
ENST00000519511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012870.hg.1 |
ENST00000519511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012870.hg.1 |
ENST00000519511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012870.hg.1 |
ENST00000519511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012870.hg.1 |
ENST00000519511.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012870.hg.1 |
ENST00000520593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012870.hg.1 |
ENST00000520593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012870.hg.1 |
ENST00000520593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012870.hg.1 |
ENST00000520593.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012870.hg.1 |
ENST00000520648.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012870.hg.1 |
ENST00000520648.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012870.hg.1 |
ENST00000520648.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012870.hg.1 |
ENST00000520648.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012870.hg.1 |
NM_003714.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012870.hg.1 |
NM_003714.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012870.hg.1 |
NM_003714.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012870.hg.1 |
NM_003714.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012870.hg.1 |
NM_003714.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012870.hg.1 |
NM_003714.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012870.hg.1 |
NM_003714.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012870.hg.1 |
NM_003714.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012870.hg.1 |
NM_003714.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012870.hg.1 |
NM_003714.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012870.hg.1 |
NM_003714.2 |
Refseq |