Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000263071.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000263071.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000263071.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000263071.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000263071.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000263071.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000263071.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000263071.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000263071.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000263071.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000263071.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000263071.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000263071.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000263071.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000263071.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000263071.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000263071.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000263071.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000263071.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000348987.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000348987.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000348987.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000348987.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000348987.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000348987.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000348987.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000348987.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000348987.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000348987.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000348987.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000348987.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000348987.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000348987.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000434376.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000434376.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000434376.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000434376.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000434376.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000434376.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000434376.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000434376.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000434376.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000434376.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000434376.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000434376.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000434376.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000434376.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000434376.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000434376.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000434376.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000434376.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000570902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000570902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000570902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000570902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000570902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000570902.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000571272.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000571272.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000571272.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000571272.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000571272.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000571272.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000571272.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000571272.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000571272.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000571272.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000571272.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000571272.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000571272.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000571272.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000571272.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000571272.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000571272.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000573852.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000573852.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000573852.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000573852.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000573852.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000573852.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000573852.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000573852.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000573852.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000573852.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000573852.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000573852.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000573852.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000573852.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000573852.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000573852.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000573852.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000573852.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000573867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000573867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000573867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000573867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000573867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000574545.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000574545.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000574545.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000576012.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000576012.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000576012.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000576012.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000576012.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000576012.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000576012.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000576012.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000576012.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000576012.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000576012.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000576012.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000576012.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000576012.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000576012.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000576012.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000576012.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000576012.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
ENST00000576012.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_003693.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_003693.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_003693.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_003693.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_003693.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_003693.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_003693.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_003693.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_003693.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_003693.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_003693.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_003693.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_003693.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_003693.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_003693.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_003693.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_003693.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_003693.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_003693.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_003693.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_145350.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_145350.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_145350.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_145350.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_145350.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_145350.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_145350.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_145350.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_145350.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_145350.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_145350.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_145350.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_145350.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_145350.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_145350.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_145350.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_145350.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_145350.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NM_145350.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_028075.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_028075.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_028075.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_028075.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_028075.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_028075.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_028075.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_028075.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_028075.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_028075.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_028075.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_028075.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_028075.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_028075.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_028075.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_028075.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_028075.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_028075.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_028075.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_102409.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_102409.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_102409.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_102409.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_102409.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_102409.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_102409.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_102409.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_102409.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_102409.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_102409.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_102409.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_102409.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_102409.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_102409.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_102409.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_102409.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_102409.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_102409.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_102409.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009392.hg.1 |
NR_102409.1 |
Refseq |