Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019375.hg.1 |
TC09001541.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019375.hg.1 |
ENST00000373887.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019375.hg.1 |
ENST00000540010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019375.hg.1 |
NM_001190945.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019375.hg.1 |
NM_005658.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
ENST00000373887.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
ENST00000373887.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
ENST00000373887.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
ENST00000373887.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
ENST00000373887.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
ENST00000373887.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
ENST00000373887.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
ENST00000373887.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
ENST00000373887.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
ENST00000373887.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
ENST00000373887.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
ENST00000373887.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
ENST00000373887.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
ENST00000373887.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
ENST00000373887.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
ENST00000540010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
ENST00000540010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
ENST00000540010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
ENST00000540010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
ENST00000540010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
ENST00000540010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
ENST00000540010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
ENST00000540010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
ENST00000540010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
ENST00000540010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
ENST00000540010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
ENST00000540010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
ENST00000540010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
ENST00000540010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
ENST00000546084.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
ENST00000546084.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
ENST00000546084.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
ENST00000546084.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
ENST00000546084.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
ENST00000546084.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
ENST00000546084.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
ENST00000546084.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
ENST00000546084.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
NM_001190945.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
NM_001190945.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
NM_001190945.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
NM_001190945.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
NM_001190945.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
NM_001190945.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
NM_001190945.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
NM_001190945.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
NM_001190945.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
NM_001190945.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
NM_001190945.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
NM_001190945.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
NM_001190945.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
NM_001190945.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
NM_001190947.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
NM_001190947.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
NM_001190947.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
NM_001190947.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
NM_001190947.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
NM_001190947.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
NM_001190947.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
NM_001190947.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
NM_001190947.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
NM_005658.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
NM_005658.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
NM_005658.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
NM_005658.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
NM_005658.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
NM_005658.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
NM_005658.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
NM_005658.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
NM_005658.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
NM_005658.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
NM_005658.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
NM_005658.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
NM_005658.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011393.hg.1 |
NM_005658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177683.hg.1 |
ENST00000373887.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900177683.hg.1 |
ENST00000540010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900177683.hg.1 |
NM_001190945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177683.hg.1 |
NM_005658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177683.hg.1 |
ENST00000373887.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900177683.hg.1 |
ENST00000540010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900177683.hg.1 |
NM_001190945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177683.hg.1 |
NM_005658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177683.hg.1 |
TC0900011393.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
ENST00000373887.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
ENST00000373887.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
ENST00000373887.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
ENST00000373887.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
ENST00000373887.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
ENST00000373887.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
ENST00000373887.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
ENST00000373887.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
ENST00000373887.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
ENST00000373887.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
ENST00000373887.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
ENST00000373887.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
ENST00000373887.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
ENST00000373887.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
ENST00000373887.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
ENST00000540010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
ENST00000540010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
ENST00000540010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
ENST00000540010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
ENST00000540010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
ENST00000540010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
ENST00000540010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
ENST00000540010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
ENST00000540010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
ENST00000540010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
ENST00000540010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
ENST00000540010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
ENST00000540010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
ENST00000540010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
ENST00000546084.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
ENST00000546084.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
ENST00000546084.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
ENST00000546084.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
ENST00000546084.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
ENST00000546084.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
ENST00000546084.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
ENST00000546084.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
ENST00000546084.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
NM_001190945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
NM_001190945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
NM_001190945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
NM_001190945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
NM_001190945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
NM_001190945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
NM_001190945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
NM_001190945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
NM_001190945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
NM_001190945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
NM_001190945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
NM_001190945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
NM_001190945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
NM_001190945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
NM_001190947.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
NM_001190947.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
NM_001190947.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
NM_001190947.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
NM_001190947.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
NM_001190947.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
NM_001190947.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
NM_001190947.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
NM_001190947.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
NM_005658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
NM_005658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
NM_005658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
NM_005658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
NM_005658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
NM_005658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
NM_005658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
NM_005658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
NM_005658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
NM_005658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
NM_005658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
NM_005658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
NM_005658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011393.hg.1 |
NM_005658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177682.hg.1 |
ENST00000373887.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900177682.hg.1 |
ENST00000540010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900177682.hg.1 |
NM_001190945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177682.hg.1 |
NM_005658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177682.hg.1 |
ENST00000373887.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900177682.hg.1 |
ENST00000540010.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900177682.hg.1 |
NM_001190945.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177682.hg.1 |
NM_005658.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177682.hg.1 |
TC0900011393.hg.1 |
Clariom D TC |