Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007513.hg.1 |
TC0X000571.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007513.hg.1 |
ENST00000371900.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007513.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008210.hg.1 |
ENST00000463626.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008210.hg.1 |
ENST00000463626.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008210.hg.1 |
ENST00000463626.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008210.hg.1 |
ENST00000598581.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008210.hg.1 |
ENST00000598581.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008210.hg.1 |
ENST00000598581.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008210.hg.1 |
ENST00000598581.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008210.hg.1 |
ENST00000598581.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008210.hg.1 |
ENST00000598581.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008210.hg.1 |
ENST00000598581.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008210.hg.1 |
ENST00000598581.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008210.hg.1 |
ENST00000598581.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008210.hg.1 |
ENST00000598581.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008210.hg.1 |
ENST00000598581.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008210.hg.1 |
ENST00000598581.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008210.hg.1 |
ENST00000598581.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008210.hg.1 |
ENST00000598581.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008210.hg.1 |
ENST00000598581.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008210.hg.1 |
ENST00000598581.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008210.hg.1 |
ENST00000598581.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008210.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008210.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008210.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008210.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008210.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008210.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008210.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008210.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008210.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008210.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008210.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008210.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008210.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008210.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008210.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008210.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008210.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008210.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008210.hg.1 |
ENST00000463626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008210.hg.1 |
ENST00000463626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008210.hg.1 |
ENST00000463626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008210.hg.1 |
ENST00000598581.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008210.hg.1 |
ENST00000598581.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008210.hg.1 |
ENST00000598581.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008210.hg.1 |
ENST00000598581.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008210.hg.1 |
ENST00000598581.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008210.hg.1 |
ENST00000598581.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008210.hg.1 |
ENST00000598581.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008210.hg.1 |
ENST00000598581.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008210.hg.1 |
ENST00000598581.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008210.hg.1 |
ENST00000598581.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008210.hg.1 |
ENST00000598581.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008210.hg.1 |
ENST00000598581.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008210.hg.1 |
ENST00000598581.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008210.hg.1 |
ENST00000598581.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008210.hg.1 |
ENST00000598581.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008210.hg.1 |
ENST00000598581.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008210.hg.1 |
ENST00000598581.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008210.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008210.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008210.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008210.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008210.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008210.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008210.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008210.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008210.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008210.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008210.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008210.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008210.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008210.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008210.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008210.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008210.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008210.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00156032.hg.1 |
ENST00000598581.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156032.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00156032.hg.1 |
ENST00000598581.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00156032.hg.1 |
NM_007231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00156032.hg.1 |
TC0X00008210.hg.1 |
Clariom D TC |