Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012437.hg.1 |
ENST00000296506.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012437.hg.1 |
ENST00000296506.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012437.hg.1 |
ENST00000296506.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012437.hg.1 |
ENST00000296506.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012437.hg.1 |
ENST00000296506.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012437.hg.1 |
ENST00000296506.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012437.hg.1 |
ENST00000296506.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012437.hg.1 |
ENST00000512188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012437.hg.1 |
ENST00000512188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012437.hg.1 |
ENST00000512188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012437.hg.1 |
ENST00000512188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012437.hg.1 |
ENST00000512188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012437.hg.1 |
ENST00000512188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012437.hg.1 |
ENST00000512188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012437.hg.1 |
ENST00000512188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012437.hg.1 |
ENST00000512188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012437.hg.1 |
ENST00000512188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012437.hg.1 |
NM_007281.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012437.hg.1 |
NM_007281.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012437.hg.1 |
NM_007281.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012437.hg.1 |
NM_007281.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012437.hg.1 |
NM_007281.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024067.hg.1 |
TC04001742.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024067.hg.1 |
ENST00000296506.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024067.hg.1 |
NM_007281.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024068.hg.1 |
TC04001742.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024068.hg.1 |
ENST00000296506.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024068.hg.1 |
NM_007281.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024065.hg.1 |
TC04001742.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024065.hg.1 |
ENST00000296506.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024065.hg.1 |
NM_007281.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185256.hg.1 |
ENST00000296506.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185256.hg.1 |
NM_007281.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185256.hg.1 |
ENST00000296506.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185256.hg.1 |
NM_007281.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185256.hg.1 |
TC0400012437.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400012437.hg.1 |
ENST00000296506.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012437.hg.1 |
ENST00000296506.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012437.hg.1 |
ENST00000296506.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012437.hg.1 |
ENST00000296506.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012437.hg.1 |
ENST00000296506.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012437.hg.1 |
ENST00000296506.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012437.hg.1 |
ENST00000296506.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012437.hg.1 |
ENST00000512188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012437.hg.1 |
ENST00000512188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012437.hg.1 |
ENST00000512188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012437.hg.1 |
ENST00000512188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012437.hg.1 |
ENST00000512188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012437.hg.1 |
ENST00000512188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012437.hg.1 |
ENST00000512188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012437.hg.1 |
ENST00000512188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012437.hg.1 |
ENST00000512188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012437.hg.1 |
ENST00000512188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012437.hg.1 |
NM_007281.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012437.hg.1 |
NM_007281.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012437.hg.1 |
NM_007281.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012437.hg.1 |
NM_007281.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012437.hg.1 |
NM_007281.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185252.hg.1 |
ENST00000296506.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185252.hg.1 |
ENST00000512188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185252.hg.1 |
NM_007281.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185252.hg.1 |
ENST00000296506.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185252.hg.1 |
ENST00000512188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185252.hg.1 |
NM_007281.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185252.hg.1 |
TC0400012437.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400185255.hg.1 |
ENST00000296506.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185255.hg.1 |
ENST00000512188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185255.hg.1 |
NM_007281.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185255.hg.1 |
ENST00000296506.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185255.hg.1 |
ENST00000512188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185255.hg.1 |
NM_007281.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185255.hg.1 |
TC0400012437.hg.1 |
Clariom D TC |