Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007339.hg.1 |
ENST00000265462.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007339.hg.1 |
ENST00000347941.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007339.hg.1 |
ENST00000352435.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007339.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007339.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007339.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007339.hg.1 |
TC11000597.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000347941.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000347941.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000347941.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000347941.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000347941.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000347941.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000352435.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000352435.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000352435.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000352435.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000352435.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000352435.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
ENST00000352435.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007948.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007338.hg.1 |
ENST00000265462.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007338.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007338.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007338.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007338.hg.1 |
TC11000597.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007340.hg.1 |
ENST00000265462.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007340.hg.1 |
ENST00000352435.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007340.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007340.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11007340.hg.1 |
TC11000597.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011560.hg.1 |
ENST00000495472.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157053.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157053.hg.1 |
ENST00000347941.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157053.hg.1 |
ENST00000352435.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157053.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157053.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157053.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157053.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157053.hg.1 |
ENST00000347941.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157053.hg.1 |
ENST00000352435.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157053.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157053.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157053.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157053.hg.1 |
TC1100007948.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100157052.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157052.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157052.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157052.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157052.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157052.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157052.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157052.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157052.hg.1 |
TC1100007948.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100157054.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157054.hg.1 |
ENST00000347941.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157054.hg.1 |
ENST00000352435.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157054.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157054.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157054.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157054.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157054.hg.1 |
ENST00000347941.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157054.hg.1 |
ENST00000352435.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157054.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157054.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157054.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157054.hg.1 |
TC1100007948.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300011560.hg.1 |
ENST00000495472.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157055.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157055.hg.1 |
ENST00000352435.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157055.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157055.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157055.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157055.hg.1 |
ENST00000352435.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100157055.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157055.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100157055.hg.1 |
TC1100007948.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000265462.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000347941.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000347941.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000347941.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000347941.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000347941.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000347941.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000352435.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000352435.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000352435.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000352435.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000352435.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000352435.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
ENST00000352435.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_012094.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_181651.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_181652.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007948.hg.1 |
NM_181652.2 |
Refseq |