Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011955.hg.1 |
ENST00000248984.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011955.hg.1 |
ENST00000486651.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011955.hg.1 |
ENST00000543051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011955.hg.1 |
NM_015372.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011955.hg.1 |
TC22000674.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008495.hg.1 |
ENST00000484682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008495.hg.1 |
ENST00000484682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008495.hg.1 |
ENST00000484682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008495.hg.1 |
ENST00000486651.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008495.hg.1 |
ENST00000486651.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008495.hg.1 |
ENST00000486651.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008495.hg.1 |
ENST00000486651.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008495.hg.1 |
ENST00000486651.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008495.hg.1 |
ENST00000623735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008495.hg.1 |
ENST00000623735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008495.hg.1 |
ENST00000623735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008495.hg.1 |
ENST00000623735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008495.hg.1 |
ENST00000623735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008495.hg.1 |
ENST00000623735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008495.hg.1 |
NM_001302819.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008495.hg.1 |
NM_001302819.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008495.hg.1 |
NM_001302819.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008495.hg.1 |
NM_001302819.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008495.hg.1 |
NM_001302819.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008495.hg.1 |
NM_001302819.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008495.hg.1 |
NM_001302820.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008495.hg.1 |
NM_001302820.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008495.hg.1 |
NM_001302820.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008495.hg.1 |
NM_001302820.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008495.hg.1 |
NM_001302820.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008495.hg.1 |
NM_015372.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008495.hg.1 |
NM_015372.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008495.hg.1 |
NM_015372.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008495.hg.1 |
NM_015372.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008495.hg.1 |
NM_015372.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008495.hg.1 |
NM_015372.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008495.hg.1 |
NR_126507.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008495.hg.1 |
NR_126507.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008495.hg.1 |
NR_126507.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008495.hg.1 |
NR_126507.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008495.hg.1 |
NR_126507.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011958.hg.1 |
ENST00000248984.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011958.hg.1 |
ENST00000486651.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011958.hg.1 |
NM_015372.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011958.hg.1 |
TC22000674.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2200161183.hg.1 |
TC2200008495.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200161183.hg.1 |
ENST00000484682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161183.hg.1 |
ENST00000486651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161183.hg.1 |
ENST00000623735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161183.hg.1 |
NM_001302819.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200161183.hg.1 |
NM_001302820.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200161183.hg.1 |
NM_015372.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200161183.hg.1 |
NR_126507.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200161183.hg.1 |
ENST00000484682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161183.hg.1 |
ENST00000486651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161183.hg.1 |
ENST00000623735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161183.hg.1 |
NM_001302819.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200161183.hg.1 |
NM_001302820.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200161183.hg.1 |
NM_015372.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200161183.hg.1 |
NR_126507.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200161185.hg.1 |
TC2200008495.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200161185.hg.1 |
ENST00000486651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161185.hg.1 |
ENST00000623735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161185.hg.1 |
NM_015372.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200161185.hg.1 |
NR_126507.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200161185.hg.1 |
ENST00000486651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161185.hg.1 |
ENST00000623735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161185.hg.1 |
NM_015372.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200161185.hg.1 |
NR_126507.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008495.hg.1 |
ENST00000484682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008495.hg.1 |
ENST00000484682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008495.hg.1 |
ENST00000484682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008495.hg.1 |
ENST00000486651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008495.hg.1 |
ENST00000486651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008495.hg.1 |
ENST00000486651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008495.hg.1 |
ENST00000486651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008495.hg.1 |
ENST00000486651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008495.hg.1 |
ENST00000623735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008495.hg.1 |
ENST00000623735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008495.hg.1 |
ENST00000623735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008495.hg.1 |
ENST00000623735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008495.hg.1 |
ENST00000623735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008495.hg.1 |
ENST00000623735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008495.hg.1 |
NM_001302819.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008495.hg.1 |
NM_001302819.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008495.hg.1 |
NM_001302819.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008495.hg.1 |
NM_001302819.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008495.hg.1 |
NM_001302819.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008495.hg.1 |
NM_001302819.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008495.hg.1 |
NM_001302820.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008495.hg.1 |
NM_001302820.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008495.hg.1 |
NM_001302820.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008495.hg.1 |
NM_001302820.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008495.hg.1 |
NM_001302820.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008495.hg.1 |
NM_015372.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008495.hg.1 |
NM_015372.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008495.hg.1 |
NM_015372.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008495.hg.1 |
NM_015372.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008495.hg.1 |
NM_015372.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008495.hg.1 |
NM_015372.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008495.hg.1 |
NR_126507.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008495.hg.1 |
NR_126507.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008495.hg.1 |
NR_126507.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008495.hg.1 |
NR_126507.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008495.hg.1 |
NR_126507.1 |
Refseq |