Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000245448.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000245448.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000245448.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000245448.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000245448.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000245448.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000245448.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000245448.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000245448.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000245448.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000245448.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000245448.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000311459.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000311459.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000311459.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000311459.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000311459.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000311459.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000311459.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000311459.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000311459.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000311459.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000311459.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000311459.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000311459.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000311459.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000311459.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000311459.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000358473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000358473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000358473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000358473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000358473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000358473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000426751.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000426751.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000426751.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000426751.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000426751.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000426751.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000426751.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000426751.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000426751.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000426751.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000426751.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000553774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000553774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000553774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000553774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000553774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000553774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000554204.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000554204.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000554204.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000554204.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000554204.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000554204.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000554204.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000554204.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000554204.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000554204.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000554204.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000554438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000554438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000554438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000554438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000554438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000554438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000554438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000554438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000554438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000554438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000554951.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000554951.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000554951.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000554951.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000554951.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000554951.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000554951.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000555939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000555939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000555939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000555939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000555939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000555939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000555939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000556962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000556962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000556962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000557421.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000557421.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000557421.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000557421.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000557421.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000557421.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
ENST00000557421.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NM_001003803.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NM_001003803.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NM_001003803.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NM_001003803.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NM_001003803.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NM_001003803.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NM_001003803.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NM_001003803.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NM_001003803.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NM_001003803.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NM_001003803.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NM_001003803.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NM_001003803.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NM_001003805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NM_001003805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NM_001003805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NM_001003805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NM_001003805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NM_001003805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NM_001003805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NM_001003805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NM_001003805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NM_001003805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NM_001003805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NM_001003805.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NM_015684.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NM_015684.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NM_015684.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NM_015684.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NM_015684.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NM_015684.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NM_015684.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NM_015684.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NM_015684.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NM_015684.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NM_015684.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NM_015684.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NM_015684.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NR_033761.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NR_033761.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NR_033761.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NR_033761.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NR_033761.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NR_033761.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NR_033761.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NR_033761.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NR_033761.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NR_033761.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NR_033761.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NR_033761.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NR_033761.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007112.hg.1 |
NR_033761.1 |
Refseq |